Merck
A potent ruthenium(II) antitumor complex bearing a lipophilic levonorgestrel group.
Inorganic chemistry (2011-08-13)
José Ruiz, Venancio Rodríguez, Natalia Cutillas, Arturo Espinosa, Michael J Hannon
RESUMEN
The novel steroidal conjugate 17-α-[2-phenylpyridyl-4-ethynyl]-19-nortestosterone (LEV-ppy) (1) and the steroid-C,N-chelate ruthenium(II) conjugate [Ru(η(6)-p-cymene)(LEV-ppy)Cl] (2) have been prepared. At 48 h incubation time, complex 2 is more active than cisplatin (about 8-fold) in T47D (breast cancer) and also shows an improved efficiency when compared to its nonsteroidal analogue [Ru(η(6)-p-cymene)(ppy)Cl] (ppy = phenylpyridine) (3) in the same cell line. The act of conjugating a levonorgestrel group to a ruthenium(II) complex resulted in synergistic effects between the metallic center and the steroidal ligand, creating highly potent ruthenium(II) complexes from the inactive components. The interaction of 2 with DNA was followed by electrophoretic mobility. Theoretical density functional theory calculations on complex 2 show the metal center far away from the lipophilic steroidal moiety and a labile Ru-Cl bond that allows easy replacement of Cl by N-nucleophiles such as 9-EtG, thus forming a stronger Ru-N bond. We also found a minimum energy location for the chloride counteranion (4(+)·Cl(-)) inside the pseudocavity formed by the α side of the steroid moiety, the phenylpyridine chelating subsystem, and the guanine ligand, i.e., a host-guest species with a rich variety of nonbonding interactions that include nonclassical C-H···anion bonds, as supported by electrospray ionization mass spectra.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
9-Ethylguanine