Merck
Quadrupole parameters of 11B in crystalline CaO.B2O3.
Solid state nuclear magnetic resonance (1992-12-01)
D Mao, P J Bray
RESUMEN
The quadrupole coupling constant (Qcc) and asymmetry parameter (eta) of 11B in crystalline CaO.B2O3 have been measured employing three different NQR and NMR methods: (1) 11B and 10B NQR; (2) 11B NQR and NMR; and (3) the 11B Zeeman NQR powder pattern. It is found that Qcc = 2594.3 +/- 0.5 kHz and eta = 0.515 +/- 0.001 at 77 K, and Qcc = 2573.5 +/- 0.5 kHz and eta = 0.511 +/- 0.002 at 300 K. These values are in agreement with, but far more accurate than, values obtained from a fourth procedure: measurement of the second-order quadrupolar effects evident in the m = + 1/2<-->m = - 1/2 transition of the 11B NMR spectrum.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Metaborato de calcio, technical, 39-44% B2O3 basis, 31-37% CaO basis, powder