Merck
  • Synthesis of o-(dimethylamino)aryl ketones, acridones, acridinium salts, and 1H-indazoles by the reaction of hydrazones and arynes.
Synthesis of o-(dimethylamino)aryl ketones, acridones, acridinium salts, and 1H-indazoles by the reaction of hydrazones and arynes.
The Journal of organic chemistry (2012-12-05)
Anton V Dubrovskiy, Richard C Larock
RESUMEN
A novel, efficient route to biologically and pharmaceutically important o-(dimethylamino)aryl ketones, acridones, acridinium salts, and 1H-indazoles has been developed starting from readily available hydrazones of aldehydes and o-(trimethylsilyl)aryl triflates. The reaction proceeds through arynes under mild conditions, tolerates a wide range of functional groups, and provides the final products in good to excellent yields.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
9(10H)-Acridanone, 99%