Merck
  • Cyclooxygenase selectivity and the risk of gastro-intestinal complications of various non-steroidal anti-inflammatory drugs: a clinical consideration.
Cyclooxygenase selectivity and the risk of gastro-intestinal complications of various non-steroidal anti-inflammatory drugs: a clinical consideration.
Inflammation research : official journal of the European Histamine Research Society ... [et al.] (1998-11-27)
S Kawai
RESUMEN
Severe gastro-intestinal complications are a major cause of NSAID-induced deaths in cases of rheumatoid arthritis. We measured COX selectivity by using an intact cell assay system, and found that NS-398 is a highly COX-2-selective inhibitor. Meloxicam, etodolac and diclofenac also showed high COX-2 selectivity. Zaltoprofen, loxoprofen-SRS (active metabolite of loxoprofen), 6-MNA (active metabolite of nabumetone) and ibuprofen showed intermediate COX-2 selectivity. The lowest COX-2 selectivities, which means the highest COX-1 selectivities, were observed in indomethacin, aspirin, and oxaprozin. There appears to be a good relationship between our data and some clinical data of severe gastro-intestinal toxicity. The more a given NSAID is selective for COX-2, the safer it is for clinical use. In conclusion, to anticipate the safety of NSAIDs, we find that an intact cell assay system, using human cells for measurement of COX selectivity, may be more useful than using direct enzyme assay systems.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Loxoprofen, solid