Merck
  • Histone H3 K36 methylation is mediated by a trans-histone methylation pathway involving an interaction between Set2 and histone H4.
Histone H3 K36 methylation is mediated by a trans-histone methylation pathway involving an interaction between Set2 and histone H4.
Genes & development (2008-10-17)
Hai-Ning Du, Ian M Fingerman, Scott D Briggs
RESUMEN
Set2-mediated H3 K36 methylation is an important histone modification on chromatin during transcription elongation. Although Set2 associates with the phosphorylated C-terminal domain (CTD) of RNA polymerase II (RNAPII), the mechanism of Set2 binding to chromatin and subsequent exertion of its methyltransferase activity is relatively uncharacterized. We identified a critical lysine residue in histone H4 that is needed for interaction with Set2 and proper H3 K36 di- and trimethylation. We also determined that the N terminus of Set2 contains a histone H4 interaction motif that allows Set2 to bind histone H4 and nucleosomes. A Set2 mutant lacking the histone H4 interaction motif is able to bind to the phosphorylated CTD of RNAPII and associate with gene-specific loci but is defective for H3 K36 di- and trimethylation. In addition, this Set2 mutant shows increased H4 acetylation and resistance to 6-Azauracil. Overall, our study defines a new interaction between Set2 and histone H4 that mediates trans-histone regulation of H3 K36 methylation, which is needed for the preventative maintenance and integrity of the genome.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
6-Azauracil, ≥98%