Merck
  • Simultaneous determination of malachite green, gentian violet and their leuco-metabolites in aquatic products by high-performance liquid chromatography-linear ion trap mass spectrometry.
Simultaneous determination of malachite green, gentian violet and their leuco-metabolites in aquatic products by high-performance liquid chromatography-linear ion trap mass spectrometry.
Journal of chromatography. A (2007-10-24)
Xueli Wu, Gong Zhang, Yongning Wu, Xiaolin Hou, Zonghui Yuan
RESUMEN
A method has been developed for the simultaneous determination of malachite green, gentian violet and their leuco-metabolites in various aquatic products using isotope dilution liquid chromatography-linear ion trap mass spectrometry without post-column oxidation. Sample was extracted with McIlvaine buffer and acetonitrile, followed by partitioning with dichloromethane, purified on basic alumina and OASIS MCX SPE column, and finally analyzed by LC-ESI-MS/MS with the select reaction monitoring (SRM) mode. Decision limits (CCalpha, alpha=0.01) and detection capability (CCbeta, beta=0.05) of the method were in the range of 0.02-0.09 and 0.04-0.13microg/kg for MG, GV, LMG and LGV in grass carp, eel, salmon, shrimp and shellfish, respectively, recoveries of MG, GV, LMG and LGV at all fortification levels (0.25-10microg/kg) were from 80.8% to 115.7%, inter-day relative standard derivations were from 1.9% to 18.4%. This method appeared suitable for the control of MG, GV, LMG and LGV residues in aquatic products.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Leucocrystal Violet