Merck
  • A method for the transient inhibition of toxicity of secretory recombinant proteins, exemplified by bacterial alkaline phosphatase. Novel protocol for problematic DNA termini dephosphorylation.
A method for the transient inhibition of toxicity of secretory recombinant proteins, exemplified by bacterial alkaline phosphatase. Novel protocol for problematic DNA termini dephosphorylation.
MethodsX (2021-08-26)
Natalia Krawczun, Marta Bielawa, Kasjan Szemiako, Beata Łubkowska, Ireneusz Sobolewski, Agnieszka Zylicz-Stachula, Piotr M Skowron
RESUMEN
Genes encoding proteins 'toxic' to recombinant host are difficult for cloning/expression and recombinant clones are unstable. Even tightly controlled inducible T7-lac, PBAD, PL, PR promoters are not totally silent in an uninduced state and thus not adequate for highly toxic proteins. An innovative approach to engineering and expression of the gene, encoding bacterial alkaline phosphatase (BAP) is proposed. The native precursor enzyme contains a signal peptide at the N-terminus and is secreted to the Escherichia coli (E. coli) periplasm. The signal peptide is then removed that allows oxidation and formation of active dimers. To decrease toxicity of the bap gene, its secretion leader coding section was replaced with a N-terminal His6-tag. The gene was expressed in E. coli in a PBAD vector, resulting in the accumulation of soluble His6-BAP in the cytoplasm. The His6-BAP was neutral to the cells, as no maturation was possible in the reducing cytoplasm. The purified homogenous protein was further reactivated in a redox buffer containing the protein structure stabilizing cofactors. The His6-BAP exhibited high activity. A dephosphorylation protocol for all types of DNA termini was developed.The method appears well suited for the industrial production of BAP and can be applied to other problematic proteins.• Efficient toxic gene expression • Novel approach to toxic gene cloning, engineering, expression, purification and reactivation of the transiently inactivated enzyme • Scaled-up production of ultrapure BAP • Improved protocol for all types of DNA termini dephosphorylation.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Comprimidos de inhibidor de proteasas SIGMAFAST, For General Use
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, ammonium sulfate suspension, 30-90 units/mg protein (modified Warburg-Christian, in glycine buffer)
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad