Merck
Anti-Inflammatory and Cytoprotective Effect of Plant Sterol and Galactooligosaccharides-Enriched Beverages in Caco-2 Cells.
Journal of agricultural and food chemistry (2019-07-11)
Gabriel López-García, Antonio Cilla, Reyes Barberá, Amparo Alegría
RESUMEN
Plant sterol (PS) (1 g/100 mL) enriched milk-based fruit beverages with or without galactooligosaccharides (GOS) (1.8 g/100 mL) were used in differentiated Caco-2 cells. Their potential cytopreventive effect against oxidative stress induced by cholesterol oxidation products (COPs) and their anti-inflammatory properties were evaluated. Preincubation (24 h) with bioaccessible fractions from beverages without and with GOS (MfB and MfB-G) completely prevented the COPs (60 μM/4 h) induced oxidative stress independent to GOS presence with exception to calcium influx and GSH content, where a partial protective effect was observed. Besides, MfB produced a significant (p < 0.05) reduction of IL-8 (40%) and IL-6 (50%) after IL-1β-induction (1 ng/mL/24 h) through the inhibition of NF-κB p65 translocation into the nucleus (16%) compared to control cells, while GOS presence compromised their anti-inflammatory effect. Therefore, PS-enriched milk-based fruit beverage could be an interesting strategy to prevent intestinal injury produced by COPs and to attenuate the pro-inflammatory process in intestinal human diseases. GOS addition had no extra beneficial antioxidant effect and even reduced their anti-inflammatory properties.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
3,3′-Dihexyloxacarbocyanine iodide, 98%
Sigma-Aldrich
Fluo-3, suitable for fluorescence, ~70%
Sigma-Aldrich
5-Cholesten-3β-ol-7-one, ≥90%