Merck
Telomeres acquire embryonic stem cell characteristics in induced pluripotent stem cells.
Cell stem cell (2009-02-10)
Rosa M Marion, Katerina Strati, Han Li, Agueda Tejera, Stefan Schoeftner, Sagrario Ortega, Manuel Serrano, Maria A Blasco
RESUMEN
Telomere shortening is associated with organismal aging. iPS cells have been recently derived from old patients; however, it is not known whether telomere chromatin acquires the same characteristics as in ES cells. We show here that telomeres are elongated in iPS cells compared to the parental differentiated cells both when using four (Oct3/4, Sox2, Klf4, cMyc) or three (Oct3/4, Sox2, Klf4) reprogramming factors and both from young and aged individuals. We demonstrate genetically that, during reprogramming, telomere elongation is usually mediated by telomerase and that iPS telomeres acquire the epigenetic marks of ES cells, including a low density of trimethylated histones H3K9 and H4K20 and increased abundance of telomere transcripts. Finally, reprogramming efficiency of cells derived from increasing generations of telomerase-deficient mice shows a dramatic decrease in iPS cell efficiency, a defect that is restored by telomerase reintroduction. Together, these results highlight the importance of telomere biology for iPS cell generation and functionality.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Anticuerpo anti-trimetil-histona H3 (Lys9), Upstate®, from rabbit
Sigma-Aldrich
Anticuerpo anti-HP1γ, clon 42s2, clone 42s2, Upstate®, from mouse