Merck
  • Estrogen receptor-based fluorescence polarization assay for bisphenol analogues and molecular modeling study of their complexation mechanism.
Estrogen receptor-based fluorescence polarization assay for bisphenol analogues and molecular modeling study of their complexation mechanism.
Analytica chimica acta (2018-08-26)
Jie Zhang, Wenfu Wu, Yongjun Wang, XiaoJia Xing, Shuning Zhong, Tianzhu Guan, Tiehua Zhang, Ligang Hou, Tiezhu Li
RESUMEN
A fluorescence polarization (FP) assay based on estrogen receptor was developed for the determination of bisphenol compounds (BPs). The human estrogen receptor α ligand binding domain (hERα-LBD) and coumestrol were employed as recognition element and fluorescent probe, respectively. Competitive displacement of tracer from receptor suggested that BPs exhibited dose-dependent binding to hERα-LBD. In order to elucidate the structural basis for the interaction between BPs and hERα-LBD, molecular dynamics simulations were performed to explore their complexation mechanism. The docked bisphenol compounds adopted agonist/antagonist conformations with varying positions and orientations in the hydrophobic binding pocket, depending on their structural characteristics of bridging moieties. Interestingly, the calculated binding energies were generally correlated with the experimentally measured affinities, indicating a potential advantage of the molecular modeling approach in predicting the binding potencies of putative ligands. Considering that the real samples may contain more than one BP, the established FP assay can potentially be used as a pre-screening method to determine the total amounts of bisphenol compounds.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
4,4′-Sulfonyldiphenol, 98%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
4,4′-Cyclohexylidenebisphenol, 98%
En este momento no podemos mostrarle ni los precios ni la disponibilidad