Merck
V900155
Sigma-Aldrich
1,4-Diazabicyclo[2.2.2]octane
Vetec, reagent grade, 98%
Sinónimos:
TED, Triethylenediamine
About This Item
Fórmula empírica (notación de Hill):
C6H12N2
Número de CAS:
Peso molecular:
112.17
Beilstein/REAXYS Number:
103618
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
grade
reagent grade
vapor pressure
2.9 mmHg ( 50 °C)
product line
Vetec
assay
98%
refractive index
n20/D 1.4634 (lit.)
mp
156-159 °C (lit.)
density
1.02 g/mL at 25 °C (lit.)
SMILES string
N12CCN(CC2)CC1
InChI
1S/C6H12N2/c1-2-8-5-3-7(1)4-6-8/h1-6H2
InChI key
IMNIMPAHZVJRPE-UHFFFAOYSA-N
Legal Information
Vetec is a trademark of Merck KGaA, Darmstadt, Germany
signalword
Danger
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Sol. 1 - Skin Irrit. 2
Storage Class
4.1B - Flammable solid hazardous materials
wgk_germany
WGK 1
flash_point_f
143.6 °F - closed cup
flash_point_c
62 °C - closed cup
Visite la Librería de documentos
Daniel Horák et al.
Colloids and surfaces. B, Biointerfaces, 149, 64-71 (2016-10-11)
Changes in cholesterol concentration in the plasma membrane of presynaptic nerve terminals nonspecifically modulate glutamate transport and homeostasis in the central nervous system. Reduction of the cholesterol content in isolated rat brain nerve terminals (synaptosomes) using cholesterol-depleting agents decreases the
Nazanin Ghazanfari et al.
The Journal of physiology, 592(13), 2881-2897 (2014-05-27)
Muscle-specific kinase (MuSK) autoantibodies from myasthenia gravis patients can block the activation of MuSK in vitro and/or reduce the postsynaptic localization of MuSK. Here we use a mouse model to examine the effects of MuSK autoantibodies upon some key components
Veronica Francardo et al.
Brain : a journal of neurology, 137(Pt 7), 1998-2014 (2014-04-24)
The sigma-1 receptor, an endoplasmic reticulum-associated molecular chaperone, is attracting great interest as a potential target for neuroprotective treatments. We provide the first evidence that pharmacological modulation of this protein produces functional neurorestoration in experimental parkinsonism. Mice with intrastriatal 6-hydroxydopamine
Sebastiaan Theuns et al.
Veterinary microbiology, 172(1-2), 23-34 (2014-05-20)
Group A rotaviruses (RVA) are an important cause of diarrhea in young piglets, resulting in significant economic losses. However, the role of RVA in the etiology of piglet diarrhea on Belgian swine farms was previously unreported. In the present study
Tania Morielli et al.
Reproduction (Cambridge, England), 149(1), 113-123 (2014-11-12)
Oxidative stress, generated by excessive reactive oxygen species (ROS) or decreased antioxidant defenses (and possibly both), is associated with male infertility. Oxidative stress results in redox-dependent protein modifications, such as tyrosine nitration and S-glutathionylation. Normozoospermic sperm samples from healthy individuals