Merck
V800032
Sigma-Aldrich
Ammonium acetate
AR, ≥98%
About This Item
Fórmula lineal:
CH3CO2NH4
Número de CAS:
Peso molecular:
77.08
Beilstein/REAXYS Number:
4186741
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
assay:
≥98%
grade:
AR
form:
solid
En este momento no podemos mostrarle ni los precios ni la disponibilidad
grade
AR
product line
Vetec
assay
≥98%
form
solid
pH
6.7-7.3 (5% in water)
mp
110-112 °C (dec.) (lit.)
anion traces
chloride (Cl-): ≤0.0005%
phosphate (PO43-): ≤0.0005%
sulfate (SO42-): ≤0.002%
cation traces
Ca: ≤0.001%
Fe: ≤0.0002%
Mg: ≤0.0005%
heavy metals (as Pb): ≤0.0002%
SMILES string
N.CC(O)=O
InChI
1S/C2H4O2.H3N/c1-2(3)4;/h1H3,(H,3,4);1H3
InChI key
USFZMSVCRYTOJT-UHFFFAOYSA-N
Legal Information
Vetec is a trademark of Merck KGaA, Darmstadt, Germany
Storage Class
11 - Combustible Solids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Visite la Librería de documentos
S Baron-Cohen et al.
Molecular psychiatry, 20(3), 369-376 (2014-06-04)
Autism affects males more than females, giving rise to the idea that the influence of steroid hormones on early fetal brain development may be one important early biological risk factor. Utilizing the Danish Historic Birth Cohort and Danish Psychiatric Central
Nazish Ahmed et al.
Tissue engineering. Part A, 20(15-16), 2224-2233 (2014-03-13)
One of the factors preventing clinical application of regenerative medicine to degenerative cartilage diseases is a suitable source of cells. Chondrocytes, the only cell type of cartilage, grown in vitro under culture conditions to expand cell numbers lose their phenotype
Martijs J Jonker et al.
Nucleic acids research, 42(11), e94-e94 (2014-04-29)
Structural variations in genomes are commonly studied by (micro)array-based comparative genomic hybridization. The data analysis methods to infer copy number variation in model organisms (human, mouse) are established. In principle, the procedures are based on signal ratios between test and
Mette Stæhr et al.
American journal of physiology. Renal physiology, 309(3), F235-F241 (2015-05-15)
In nephrotic syndrome, aberrant glomerular filtration of plasminogen and conversion to active plasmin in preurine are thought to activate proteolytically epithelial sodium channel (ENaC) and contribute to sodium retention and edema. The ENaC blocker amiloride is an off-target inhibitor of
Lucie Nováková et al.
Analytica chimica acta, 853, 637-646 (2014-12-04)
The conditions for the analysis of selected doping substances by UHPSFC-MS/MS were optimized to ensure suitable peak shapes and maximized MS responses. A representative mixture of 31 acidic and basic doping agents was analyzed, in both ESI+ and ESI- modes.