Documentos clave
59477
Optimize HPLC Pump Part for Spectra-Physics
for use with 8800, 8810, Isochrom, P-Series, Spectra Physics A3102-010
About This Item
Código UNSPSC:
41115854
envase
pkg of 1
fabricante / nombre comercial
Spectra Physics A3102-010
compatibilidad
for use with 8800, 8810, Isochrom, P-Series
Características y beneficios
  • Offers optimal concentricity.        
  • Specifically cut in relation to the grain of the crystal lattice.         Maximal resistance to breakage.        
  • Superior wear characteristics.       
  • Proprietary attachment of the rod to the piston ferrule ensures concentricity.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)