Documentos clave
57625
Sample Loop for Model 7725 Injector
volume 20 μL
About This Item
Código UNSPSC:
41115854
En este momento no podemos mostrarle ni los precios ni la disponibilidad
volumen
20 μL
Descripción general
Sample loop for model 7725 injector (volume 20 μL) is designed for advanced fluid-handling solutions for a variety of analytical instrumentation and in vitro diagnostic systems. It can be employed in high performance liquid chromatographic (HPLC) systems and is quite accurate and precise in injecting 1 μL-5 mL concentration of samples.
Productos recomendados
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Visite la Librería de documentos
Artículos
Correct common HPLC issues to restore column performance, addressing peak problems, retention time, noise, and resolution with chromatograms.