Merck
23393-U
Supelco
Washed Stainless Steel Clip for ATD-400 Glass TDU, pk 100
greener alternative
About This Item
Código UNSPSC:
41104008
NACRES:
NB.24
envase
pack of 100 ea
Nivel de calidad
características de los productos alternativos más sostenibles
Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
técnicas
active air sampling: suitable
aplicaciones
air monitoring
environmental
industrial hygiene
categoría alternativa más sostenible
Descripción general
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. Thermal desorption tubes are reusable at least 50 to 100 times, and reduces the usage of accessories like transfer vials, caps, and pipette tips, thus aligns with “Waste Prevention”. Also, these tubes eliminate the usage of toxic solvents like CS2 to extract the VOCs of interest, and thus aligns with “Safer Solvents and Auxiliaries” principle. Click here for more information.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)