Documentos clave
22434
Mirror with Rotating Head
chrome-plated handle, pkg of 1 ea
About This Item
Código UNSPSC:
41100000
Materiales
chrome-plated handle
envase
pkg of 1 ea
Descripción general
Ideal for examining inside of injection and detector fittings for ferrule or column fragments. Rotating head allows wide range of movement.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)