Documentos clave
20448
Ratchet Wrench
wrench size 9/16 in.
About This Item
Código UNSPSC:
41100000
tamaño de la llave
9/16 in.
compatibilidad
for use with Swagelok 1/4 in. fittings
Descripción general
Gets into tight places. Jaws open to slip around the nut, then close. Convenient to use in a small chromatograph oven or other confined places. 7/16 in. wrench is for 1/8 in. Swagelok fittings, 9/16 in. wrench is for 1/4 in. Swagelok fittings.
Información legal
Swagelok is a registered trademark of Swagelok Company
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)