Saltar al contenido
Merck
Todas las fotos(11)

Documentos clave

ZRB2394

Sigma-Aldrich

Anti-eIF4G1 Antibody, clone 2C6 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sinónimos:

Eukaryotic translation initiation factor 4 gamma 1, eIF-4-gamma 1, eIF-4G 1, eIF-4G1, p220)

Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización


About This Item

Código UNSPSC:
12352203
NACRES:
NA.43

origen biológico

rabbit

Nivel de calidad

recombinante

expressed in HEK 293 cells

conjugado

unconjugated

forma del anticuerpo

purified antibody

tipo de anticuerpo

primary antibodies

clon

2C6, recombinant monoclonal

descripción

recombinant, expressed in HEK 293 cells

Línea del producto

ZooMAb® learn more

Formulario

lyophilized

purificado por

using Protein A

reactividad de especies

human, rat, mouse

envase

antibody small pack of 25 μL

características de los productos alternativos más sostenibles

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

validación mejorada

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

técnicas

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotipo

IgG

secuencia del epítopo

Internal

Nº de acceso Protein ID

Nº de acceso UniProt

categoría alternativa más sostenible

Condiciones de envío

ambient

temp. de almacenamiento

2-8°C

Información sobre el gen

human ... EIF4G1(1981)

Descripción general

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Especificidad

Clone 2C6 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects eIF4G1. It targets an epitope within 21 amino acids from the internal region and detects multiple isoforms.

Inmunógeno

KLH-conjugated linear peptide corresponding to 21 amino acids from the internal region of human Eukaryotic translation initiation factor 4 gamma 1 (eIF4G1).

Aplicación

Quality Control Testing

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected eIF4G1 in HeLa cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected eIF4G1 in PC12 cell lysate.

Affinity Binding Assay: A representative lot of this antibody bound eIF4G1 peptide with a KD of 3.9 x 10-8 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected eIF4G1 in A431, HeLa, HUVEC, and NIH3T3 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected eIF4G1 in human cerebral cortex tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Descripción de destino

Eukaryotic translation initiation factor 4 gamma 1 (UniProt: Q04637; also known as eIF-4-gamma 1, eIF-4G 1, eIF-4G1, p220) is encoded by the EIF4G1 (also known as EIF4F, EIF4G, EIF4GI) gene (Gene ID: 1981) in human. eIF4G1 is a component of the eIF4F complex that is involved in the recognition of the mRNA cap, ATP-dependent unwinding of 5′-terminal secondary structure, and recruitment of mRNA to the ribosome. It serves as a scaffold for assembly of components of EIF4F complex that starts cap-dependent translation.  This complex also plays a role in oncogenesis. Increased expression of EIF4G1 has been linked to lower median survival of patients with multiple cancers. Knockdown of eIF4G1 in prostate cancer cells is reported to reduce cyclin D1 and p-Rb protein levels and cause cell cycle delay and reduce cell viability, proliferation, and migration. eIF4G1 serves as a substrate for PKCα and its phosphorylation at serine 1186 induces its binding to mitogen-activated protein kinase–interacting kinase 1 (Mnk1). Eight isoforms of eIF4G1 have been described that are produced by alternative splicing. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, Affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Jaiswal, PK., et al. (2019). Cancer Cell Int. 19; Article 224; Jaiswal, PK., et al. (2018). Sci. Rep. 8; Article 7459; Dobrikov, M., et al. (2011). Mol. Cell. Biol. 31(14); 2947-2959).

Forma física

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Almacenamiento y estabilidad

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Otras notas

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Información legal

Affinity is a trademark of Mine Safety Appliances Co.
ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Cláusula de descargo de responsabilidad

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

¿No encuentra el producto adecuado?  

Pruebe nuestro Herramienta de selección de productos.

Código de clase de almacenamiento

11 - Combustible Solids

Clase de riesgo para el agua (WGK)

WGK 1

Punto de inflamabilidad (°F)

Not applicable

Punto de inflamabilidad (°C)

Not applicable


Elija entre una de las versiones más recientes:

Certificados de análisis (COA)

Lot/Batch Number

¿No ve la versión correcta?

Si necesita una versión concreta, puede buscar un certificado específico por el número de lote.

¿Ya tiene este producto?

Encuentre la documentación para los productos que ha comprado recientemente en la Biblioteca de documentos.

Visite la Librería de documentos

Nuestro equipo de científicos tiene experiencia en todas las áreas de investigación: Ciencias de la vida, Ciencia de los materiales, Síntesis química, Cromatografía, Analítica y muchas otras.

Póngase en contacto con el Servicio técnico