Documentos clave
Z741608
Plus Safe-Cone Filter
diam. 6.73 mm, polyethylene
Seleccione un Tamaño
50 EA
US$ 57,50
US$ 57,50
Fecha estimada de envío10 de mayo de 2025
Seleccione un Tamaño
Cambiar Vistas
50 EA
US$ 57,50
About This Item
Código UNSPSC:
41121500
NACRES:
NB.01
US$ 57,50
Fecha estimada de envío10 de mayo de 2025
Materiales
polyethylene
envase
package of 50 (individually wrapped)
fabricante / nombre comercial
Sartorius 721015
Diámetro
6.73 mm
Descripción general
The unique and replaceable polyethylene (PE) Safe-Cone Filters act as a final barrier to prevent any fluids and liquid vapours from reaching the internal components of the pipette.Standard Safe-Cone Filters are for general applications. They can be used for the same type of work as the Standard Filter is recommended for, but need to be changed less frequently. Self-sealing Plus Safe-Cone Filters are meant for applications such as radioactive work, cell culture, bacterial and virological work and molecular biology.The interval of changing the filter depends on the application and the sample. However, according to studies, the filter should be changed daily (or after 50 to 250 pipettings) and immediately in case of over-aspiration. To ensure safety of the user, forceps should be used to change the filter. The mLINEmLINE® pipettes also feature a built-in filter ejector. In addition, clean the tip cone with ethanol (70%) prior to assembling a new filter.The unique and replaceable polyethylene (PE) Safe-Cone Filters act as a final barrier to prevent any fluids and liquid vapours from reaching the internal components of the pipette.Standard Safe-Cone Filters are for general applications. They can be used for the same type of work as the Standard Filter is recommended for, but need to be changed less frequently. Self-sealing Plus Safe-Cone Filters are meant for applications such as radioactive work, cell culture, bacterial and virological work and molecular biology.The interval of changing the filter depends on the application and the sample. However, according to studies, the filter should be changed daily (or after 50 to 250 pipettings) and immediately in case of over-aspiration. To ensure safety of the user, forceps should be used to change the filter. The mLINE® pipettes also feature a built-in filter ejector. In addition, clean the tip cone with ethanol (70%) prior to assembling a new filter.
Información legal
mLINE is a registered trademark of Sartorius Biohit Liquid Handling Oy
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)