Documentos clave
Z659479
Bandelin SONOREX Ultrasonic bath accessories
insert basket for Z659460
Seleccione un Tamaño
1 EA
US$ 60,10
US$ 60,10
PRECIOS SIN IMPUESTOS NACIONALES
Check Cart for Availability
Seleccione un Tamaño
Cambiar Vistas
1 EA
US$ 60,10
About This Item
Código UNSPSC:
41103706
NACRES:
NB.74
US$ 60,10
PRECIOS SIN IMPUESTOS NACIONALES
Check Cart for Availability
Materiales
stainless steel
envase
pack of 1 ea
fabricante / nombre comercial
Bandelin K 08
disponibilidad
not available in USA and Canada
L × An × P
170 mm × 65 mm × 60 mm
Descripción general
During ultrasonic operation, nothing may be placed on the tank floor, as this could cause abrasion between the sonication material and the ultrasonic oscillating tank. The use of a suitable basket to hold the sonication material is therefore recommended. This stainless steel baskets is designed so that the sonication material is always in the optimal area of ​​the cavitation field (1-2 cm from the floor) in the oscillating tank. With the help of a basket, the sonication material can also be conveniently placed in the device and removed after operation, e.g. for rinsing, without the user coming into contact with the sonication liquid in the oscillating tank.
Características y beneficios
  • Perfect positioning in the sound field
  • Optimal loading and unloading
Información legal
SONOREX is a trademark of Bandelin Electronic GmbH & Co. KG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)