Documentos
Z649732
Techne® hybridization tube holder
for Model HB-1D, capacity, 50-mL tubes
About This Item
UNSPSC Code:
12352202
NACRES:
NB.22
feature
capacity 50-mL tubes
manufacturer/tradename
Techne F50ML1TH
availability
available only in EU
Compatibility
  • Carrier with clips for 8 × 50mL tubes
Legal Information
Techne is a registered trademark of Techne, Inc.