Documentos clave
Z324957
BRAND® BLAUBRAND® bulb pipette, calibrated to deliver (TD, EX)
capacity 4 mL, one mark
About This Item
Código UNSPSC:
41121500
NACRES:
NB.01
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Materiales
AR-Glas®
descripción
total delivery Ex + 15 sec waiting time
Características
class AS
envase
pack of 6 ea
fabricante / nombre comercial
BRAND 29706
capacidad
4 mL
Categorías relacionadas
Descripción general
Additional to DIN
All BLAUBRAND bulb pipettes are supplied with a batch number and an accompanying batch certificate in the original packaging.
According to DIN EN ISO 648.
Otras notas
BR29706 (India)
Información legal
AR-Glas is a registered trademark of Schott AG
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Visite la Librería de documentos
Protocolos
This page contains detailed information on working with and handling pipettes and pipetting aids.