Documentos clave
T4541
Supelco
Glass tubes for tube gels
I.D. × L 0.5 mm × 7.5 cm
About This Item
Código UNSPSC:
12352200
D.I. × L
0.5 mm × 7.5 cm
Características y beneficios
Although the tubes listed below have different inside diameters, they all have the same 3.0 mm outside diameter. This makes the tubes less fragile and easier to handle.
Compatibilidad
The 7.5 cm tubes fit the mini electrophoresis unit, and the 13.5 cm tubes fit the standard-size unit.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)