Merck
S8061
Sigma-Aldrich
SB 242084 dihydrochloride hydrate
≥98% (HPLC), powder
Sinónimos:
6-Chloro-2,3-dihydro-5-methyl-N-[6-[(2-methyl-3-pyridinyl)oxy]-3-pyridinyl]-1H-indole-1-carboxyamide dihydrochloride hydrate, 6-Chloro-5-methyl-1-[[2-(2-methylpyrid-3-yloxy)pyrid-5-yl]carbamoyl]indoline dihydrochloride hydrate
About This Item
Fórmula empírica (notación de Hill):
C21H19ClN4O2 · 2HCl · xH2O
Número de CAS:
Peso molecular:
467.78 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
assay
≥98% (HPLC)
form
powder
storage condition
desiccated
color
white to beige
solubility
DMSO: 15 mg/mL, clear
H2O: 5 mg/mL, clear
originator
GlaxoSmithKline
storage temp.
2-8°C
SMILES string
O.Cl.Cl.Cc1cc2CCN(C(=O)Nc3ccc(Oc4cccnc4C)nc3)c2cc1Cl
InChI
1S/C21H19ClN4O2.2ClH.H2O/c1-13-10-15-7-9-26(18(15)11-17(13)22)21(27)25-16-5-6-20(24-12-16)28-19-4-3-8-23-14(19)2;;;/h3-6,8,10-12H,7,9H2,1-2H3,(H,25,27);2*1H;1H2
InChI key
SUOZHMLTMKJQNG-UHFFFAOYSA-N
Application
SB 242084 dihydrochloride hydrate was used to study the role of 5-HT2C on food intake behavior of rats.4,5
Biochem/physiol Actions
SB 242084 is a potent and selective antagonist of 5-HT2C receptor. It increases the basal activity of dopaminergic neurons and affects the behavioral responses mediated by 5-HT2C such as mesolimbic neuron activity and food intake.3,4,5
Features and Benefits
This compound was developed by GlaxoSmithKline. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.
Caution
Hygroscopic
Legal Information
Manufactured and sold with the permission of GlaxoSmithKline Pharmaceuticals
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Visite la Librería de documentos
Claudia von Meyenburg et al.
Pharmacology, biochemistry, and behavior, 74(2), 505-512 (2002-12-14)
Rats consistently reduce their food intake following injections of bacterial lipopolysaccharides (LPS). Because inhibition of serotonergic (5-HT) activity by 8-OH-DPAT (5-HT(1A) activation) attenuates LPS-induced anorexia, we conducted a series of studies to examine whether other 5-HT-receptors are involved in the
G A Kennett et al.
Neuropharmacology, 36(4-5), 609-620 (1997-04-01)
SB 242084 has a high affinity (pKi 9.0) for the cloned human 5-HT2C receptor and 100- and 158-fold selectivity over the closely related cloned human 5-HT2B and 5-HT2A subtypes respectively. SB 242084 had over 100-fold selectivity over a range of
V Di Matteo et al.
Neuropharmacology, 38(8), 1195-1205 (1999-08-26)
Electrophysiological techniques and in vivo microdialysis were used to investigate the effect of SB 242084, a potent and selective 5-HT2C receptor antagonist in the control of nigro-striatal and mesolimbic dopaminergic function. Thus, extracellular single unit recordings were performed from neurochemically-identified
6-Chloro-5-methyl-1-[[2-[(2-methyl-3-pyridyl)oxy]-5-pyridyl]carbamoyl]- indoline (SB-242084): the first selective and brain penetrant 5-HT2C receptor antagonist.
S M Bromidge et al.
Journal of medicinal chemistry, 40(22), 3494-3496 (1997-11-14)
Brigitte S Kopf et al.
Brain research, 1306, 77-84 (2009-09-29)
LPS, a potent activator of the innate immune system, is commonly used to investigate the acute phase response to infection, including anorexia. Serotonin 2C-receptor signaling has been shown to be involved in the mediation of LPS anorexia. Here we used