Spend $ 530.000 — Save 5%; Spend $ 907.000 — Save 10%; Spend $ 1.500.000 — Save 22%*
Iniciar sesión para comprobar la elegibilidad. Comprar productos subvencionables
Seleccione un Tamaño
| Talla/SKU | Disponibilidad | Precio |
|---|---|---|
4 ea | Comprobar disponibilidad del carrito | US$ 65,70 |
Acerca de este artículo
US$ 65,70
Ahorre hasta 22%
material
round white polypropylene
feature
to hold 20 x 1,5/2.0 mL tubes
packaging
pack of 4 ea
manufacturer/tradename
Heathrow Scientific HS2134A
General description
1 of 1
Este artículo | |||
|---|---|---|---|
| description to hold, 20 x 1,5/2.0 mL tubes, round white polypropylene | description to hold, 16 x 1,5/2.0 mL tubes, square white polypropylene | description Holds 28 x 1.5-2 mL tubes, polypropylene | description green |
| manufacturer/tradename Heathrow Scientific HS2134A | manufacturer/tradename Heathrow Scientific HS2132A | manufacturer/tradename Bel-Art F18742-0001 | manufacturer/tradename Heathrow Scientific, Heathrow Scientific HS29022C |
| feature to hold 20 x 1,5/2.0 mL tubes | feature to hold 16 x 1,5/2.0 mL tubes | feature Holds 28 x 1.5-2 mL tubes | feature Holds 4 x 50 mL tubes, Holds 32 x 1.5-2 mL tubes, Holds 32 x 0.5 mL tubes, Holds 12 x 15 mL tubes |
| material round white polypropylene | material square white polypropylene | material polypropylene | material polypropylene (heavy duty) |
| packaging pack of 4 ea | packaging pack of 4 ea | packaging pack of 1 ea | packaging - |
Still not finding the right product?
Explore all of our products under Floating Microtube Rack
Elija entre una de las versiones más recientes:
¿Ya tiene este producto?