Documentos clave
HS120579
Premium Plus Slide Box
to hold, 100 slide(s), translucent red acrylonitrile-butadiene-styrene (ABS)
Seleccione un Tamaño
5 EACH
US$ 109,00
US$ 109,00
PRECIOS SIN IMPUESTOS NACIONALES
Check Cart for Availability
Seleccione un Tamaño
Cambiar Vistas
5 EACH
US$ 109,00
About This Item
Código UNSPSC:
41122800
NACRES:
NB.16
US$ 109,00
PRECIOS SIN IMPUESTOS NACIONALES
Check Cart for Availability
Materiales
polypropylene
translucent red acrylonitrile-butadiene-styrene (ABS)
Características
to hold 100 slide(s)
envase
pack of HS120579
fabricante / nombre comercial
Heathrow Scientific 120579
L × An × Al
208.3 mm × 195.6 mm × 33 mm
Descripción general
The Premium Plus Slide Box′s modern design for daily use is stylish with a wave surface texture. The slide box is easy to hold,increasing safety of sample transport with an EVA foam lined bottom of the box helping to hold slides securely in placeand protect against damage. Its unique side storage can accommodate a pen, desiccant or other small items needed for working.
Características y beneficios
  • Made from ABS and Polypropylene makes the boxes robust and eliminates potential rusting of metal parts
  • Inventory card is removable and replaceable for trouble-free writing on flat surfaces.
  • Hinged lid smoothly opens 180° sitting flat on surfaces and prevents misplacement
  • EVA foam lines the bottom of the box helping to hold slides securely in place and protect against damage
  • Multiple labeling areas, a large area on the lid of the box with "project" and "date" molded in and the back of the index card can be used as the lid is transparent
  • Vibrant translucent colors are easy to identify and enable coding options
  • Stylish wave surface texture is easy to hold, increasing safety of sample transport
  • Stackable, molded feet allow secure horizontal stacking of multiple units for safe transport or storing
  • Flexible storage, the units can be stored vertically or horizontally helping to efficiently use space available
  • Unique side storage can accommodate a pen, desiccant or other small items needed for working
  • Holds 100 - 26 x 76 mm slides
  • Patent Pending
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)