Documentos
EP5895201008
Rotor for Eppendorf® 5910 R Centrifuge
to hold, 4 x universal buckets
About This Item
UNSPSC Code:
41103909
NACRES:
NB.82
feature
to hold 4 x universal buckets
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5895201008
parameter
4,500 max. RCF (4,347 xg)
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5910R
General description
Rotor for Eppendorf(R)5910R Centrifuge
Legal Information
Eppendorf is a registered trademark of Eppendorf AG