EP5804751000
Eppendorf® Adapter For 100 mL Rectangular Bucket
holds standard, Vacutainer,Falcon and micro test tubes 1.5-2 mL, pack of 2 ea
About This Item
feature
holds standard, Vacutainer,Falcon and micro test tubes 1.5-2 mL
packaging
pack of 2 ea
manufacturer/tradename
Eppendorf® 5804751000
technique(s)
centrifugation: suitable
compatibility
for use with 100 mL retangular bucket
General description
Tube adapters for use with Eppendorf 100 mL rectangular bucket.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)