Documentos clave
EP5804738004
Eppendorf® Adapter
holds 3 x 2.6-7ml tubes/ 75 mm, pack of 2 ea
About This Item
UNSPSC Code:
41103911
NACRES:
NB.82
feature
holds 3 x 2.6-7ml tubes/ 75 mm
packaging
pack of 2 ea
manufacturer/tradename
Eppendorf® 5804738004
technique(s)
centrifugation: suitable
compatibility
for use with rotor F-34-6-38
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)