Key Documents
EP5452720008
Eppendorf® Rotor F45-12-11
for 5453 plus
About This Item
UNSPSC Code:
41103909
NACRES:
NB.82
feature
to hold 12 x 1.5/2.0 mL tubes
for 5453 plus
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5452720008
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models MiniSpin/MiniSpin Plus
Legal Information
Eppendorf is a registered trademark of Eppendorf AG