Documentos clave
EP5368000010
Eppendorf® Exchangeable Thermoblock
Holds for 4 x slides
About This Item
Código UNSPSC:
41102404
Características
Holds for 4 x slides
envase
pkg of 4 ea
fabricante / nombre comercial
Eppendorf® 5368000010
Descripción general
  • Ideal for hybridization experiments on slides, e.g., Microarrays, FISH and CISH.
  • Increased hybridization efficiency due to rapid and homogeneous mixing of the sample (when using the Thermomixer comfort).
  • Uniform heating and atmospheric control improves signal-to-background ratio, increases signal intensity and results in excellent reproducibility.
  • High level temperature stability during hybridization.
  • System ensures the highest level of homogeneity of the hybridization parameters between the chambers.
  • Reservoir pads ensure high air humidity.
  • Light-proof interior.
Product Information
For use with ThermoStat plus (heating/ cooling).
The exchangeable thermoblock system for the Eppendorf ThermoMixer® comfort and ThermoStat plus ensures a precise “fit” for all common lab tubes: from simple temperature control of a chemical to complex enzyme reactions and hybridization experiments, and from MTPs to 50 mL conical tubes – the application possibilities are virtually endless!
Features
  • A wide range of exchangeable thermoblocks to cover tubes from 0.2 mL to 50 mL as well as plates and glass slides
Heat insulated thermoblocks for ergonomic and safe operation (no direct contact with hot surfaces - you won′t burn your fingers)
  • Dry incubation technology eliminates contamination risk associated with classical waterbath incubation
  • Simple replacement of thermoblocks
  • Tubes are easy accessible
  • Whisper quiet operation and optimal temperature transfer due to complete enclosure and custom fit of tubes in the exchangeable thermoblocks
Información legal
Eppendorf is a registered trademark of Eppendorf AG
Thermomixer is a registered trademark of Eppendorf AG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)