Documentos clave
EP4861000112
Eppendorf® Xplorer® 12-channel pipettor
volume 0.5-10 μL, multifunction rocker switch, medium gray, for pipette tips 20 ul
About This Item
Código UNSPSC:
41121500
NACRES:
NB.01
Características
for pipette tips 20 ul
multifunction rocker switch
envase
case of 1 ea
fabricante / nombre comercial
Eppendorf® 4861000112
volumen
0.5-10 μL
color
medium gray
Descripción general
  • Multi-functional rocker with up is up and down is down functionality
  • Selection dial for quick and easy selection of functions
  • Self-explanatory color display without submenus and with ergonomic angle for optimal readability in every position
  • Spring-loaded tip cone for all pipettes up to 1,000 μL for minimal tip attachment forces helps to reduce stress
  • Powerful rechargeable battery for up to 8 hours work without the need for recharging
  • Separate power socket: Work can continue while charging
  • Adjustment to external conditions (e.g. geographic height, liquids with high density)
  • Lower part of pipette can be autoclaved to ensure decontamination
  • Spring-loaded tip cone (not for 1,200?μL multi-channel)
  • Xplorer plus only: Individual programming, fixed volume settings, password protection, on/off switch, additional applications, display of service intervals
Eppendorf®, a renowned life science company, specializes in developing and selling instruments, consumables, and services for liquid handling, sample handling, and cell handling in laboratories worldwide. With a diverse product range, Eppendorf products are extensively utilized in both academic and commercial researchlaboratories.
Características y beneficios
  • Electronic air-cushion pipette that offers exceptional simplicity, precision, and reproducibility.
  • Pipette is designed for easy use with a multi-function rocker that follows the logic of "up is up" for aspiration and "down is down" for dispensing.
  • The selection dial helps in choosing different modes quickly.
  • Has a self-explanatory color display that eliminates the need for submenus and is ergonomically positioned at an optimal angle, ensuring optimal readability from any position.
  • Spring-loaded tip cone on all pipettes up to 1,000 µL, minimizes tip attachment forces and helps reduce stress during pipetting.
  • Powerful rechargeable battery allows up to 8 hours of uninterrupted operation without the need for recharging.
  • The separate power socket allows it to continue working while charging the device.
  • Gets easily adjusted to different external conditions, including changes in geographic height or handling liquids with high density.
  • Autoclave the lower part of pipette to ensure decontamination.
  • The tip cone of the pipette is equipped with a spring mechanism (excluding the 1,200 µL multi-channel model).
  • Xplorer plus offers features like individual programming, fixed volume settings, password protection, an on/off switch, additional applications, and displaying service intervals.
  • Ensuring results you can rely on: Accuracy and precision consistently within limits of ISO 8655
  • You always have the choice to work with other manu- facturer’s tips: Calibrated for use with epT.I.P.S. (384) pipette tips but featuring a universal cone system
  • Misaligned pipette tips or tips falling off, or not being ejected properly are a thing of the past: Spring-loaded tip cones1 enable effortless tip attachment and SOFT eject technology reduces tip ejection forces by up to 50%
  • Making your lab work safer and helping to keep your samples free from contamination: Our multi-channel pipettes are easy and fast to dismantle and always autoclavable
  • Built to last and serve you for a long time in the lab: Premium materials and UV resistance are staples of our pipette development
  • Supporting healthy working conditions in the lab: Low weight, an ideal hand balance and low operation forces in line with the Eppendorf PhysioCare Concept
Información legal
Eppendorf is a registered trademark of Eppendorf AG
Xplorer is a registered trademark of Eppendorf AG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)