Documentos
EP0030075030
Eppendorf® epT.I.P.S.® G Racks, Biopur®
size (2-200 μL), L (53 mm), yellow, pack of 480 ea (5 trays of 96 tips)
About This Item
UNSPSC Code:
41121600
material
yellow (Tray)
sterility
sterile
feature
DNase free
packaging
pack of 480 ea (5 trays of 96 tips)
manufacturer/tradename
Eppendorf® 0030075030
L
(53 mm)
maximum volume
200 μL
size
(2-200 μL)
color
yellow
Legal Information
Biopur is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
epT.I.P.S. is a registered trademark of Eppendorf AG