Seleccione un Tamaño
| Talla/SKU | Disponibilidad | Precio |
|---|
Acerca de este artículo
material
cap (fill)
sterility
sterile
packaging
case of 4
manufacturer/tradename
Corning 3333
General description
Features and Benefits
- Consists of 1/4 flexible tubing that is heat sealable and chemically resistant
- Threaded cap made up of polyethylene
- Gamma irradiation sterilized
Legal Information
1 of 1
Este artículo | |||
|---|---|---|---|
| description sterile, 6.4 mm I.D. tubing, 70 cm length, male MPC coupling with female end cap, cap (fill), case of 4 | description tubing I.D. 9.5 mm, L 7 cm, filter size 37 mm , hydrophobic glass fiber, sterile, cap (vent), case of 4 | description sterile, fill cap, female MPC coupling, 6.4 mm I.D. barbed fitting with male end cap, cap (fill) | description I.D. 9.5 mm, size 7.94 mm , barbed fitting, sterile, cap (fill), case of 5 |
| sterility sterile | sterility sterile | sterility sterile | sterility sterile |
| manufacturer/tradename Corning 3333 | manufacturer/tradename Corning 3284 | manufacturer/tradename Corning 3328 | manufacturer/tradename Corning 3283 |
| material cap (fill) | material cap (vent) | material cap (fill) | material cap (fill) |
| packaging case of 4 | packaging case of 4 | packaging - | packaging case of 5 |
Still not finding the right product?
Explore all of our products under Corning® CellSTACK® accessories
Elija entre una de las versiones más recientes:
¿Ya tiene este producto?