Documentos clave
BR29963
BRAND® BLAUBRAND® bulb pipette with serialized numbers, calibrated to deliver (TD, EX)
capacity 15 mL, one mark
About This Item
Código UNSPSC:
41121510
Materiales
AR-Glass
Características
supplementary to DIN range
envase
pkg of 12 ea
fabricante / nombre comercial
BRAND 27452
capacidad
15 mL
Descripción general
All BLAUBRAND bulb pipettes are supplied with a batch number and an accompanying batch certificate in the original packaging.According to DIN EN ISO 648.
Información legal
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)