Documentos clave
AXYTE1004BLRS
MAXYMum Recovery® pipette tips for Eppendorf® and Socorex pipettors
capacity 1000 μL, blue, sterile
Sinónimos:
pipet tips
About This Item
UNSPSC Code:
41121600
NACRES:
NB.02
capacity:
1000 μL
sterility:
γ-irradiated
sterile
feature:
autoclavable
packaging:
case of 3456 tips racked
manufacturer/tradename:
Axygen Inc
Corning
En este momento no podemos mostrarle ni los precios ni la disponibilidad
material
blue
sterility
γ-irradiated
sterile
feature
autoclavable
packaging
case of 3456 tips racked
manufacturer/tradename
Axygen Inc
Corning
capacity
1000 μL
Other Notes
Browse all pipette tips
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Maxymum Recovery is a registered trademark of Corning, Inc.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)