Documentos
99115
Sigma-Aldrich
Outlet valve No 70691
About This Item
UNSPSC Code:
41100000
NACRES:
NB.51
General description
Control valves for cylinders in stainless steel, for corrosive gases, angled, with nut at inlet and unscrewable hose outlet.
Inlet: G5/8″ ; Hose oulet: G1/4″ (M)
Choose from one of the most recent versions:
Certificados de análisis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Atención al cliente