Key Documents
414001PLATER
Stylus (1 mL)
for use with easySpiral (platers), pkg of 1 sets
About This Item
UNSPSC Code:
41122409
NACRES:
NB.24
material
plastic bottle
packaging
pkg of 1 sets
application(s)
cosmetics
food and beverages
pharmaceutical
compatibility
for use with easySpiral (platers)
General description
One Stylus (1mL) kit with connectors for easySpiral® plater
Application
Culture media plating
Legal Information
easySpiral is a registered trademark of InterLab
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)