27383
Conjuntos de émbolo Hamilton® con puntas para las gamas 1000, 1700 y 1800
for use with Hamilton 1801, N, RN, pkg of 1 ea
About This Item
material
PTFE plunger tip
size
10 μL
feature
1801N, 1801RN
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 32193
compatibility
for use with Hamilton 1801, N, RN
General description
Para reparaciones en campo de las series 1000, 1700, y 1800. No válidads para jeringas Sample Lock.
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificados de análisis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Atención al cliente