SPKGENELC044
Spare part, filter mat
W × H × D 89 mm × 89 mm × 10 mm
About This Item
Código UNSPSC:
23151806
Productos recomendados
Materiales
fibrous
Nivel de calidad
Línea del producto
Mobius®
An × Al × P
89 mm × 89 mm × 10 mm
compatibilidad
for use with Mobius®
Categorías relacionadas
Descripción general
These filters are used on BioProcessing systems to capture dust, dirt, and other airborne particles. They ensure optimal performance and of the equipment.
This spare part is required for the preventive maintenance service of some of our BioProcessing systems, it is recommended to have one in stock and to change it every 12 months.
This spare part is required for the preventive maintenance service of some of our BioProcessing systems, it is recommended to have one in stock and to change it every 12 months.
Envase
Pack of 5 units
Información legal
MOBIUS is a registered trademark of Merck KGaA, Darmstadt, Germany