Saltar al contenido
Merck
Todas las fotos(1)

Documentos clave

MABC1689

Sigma-Aldrich

Anti-GFP, clone 2B6

new
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización


About This Item

Código UNSPSC:
12352203
eCl@ss:
32160702
En este momento no podemos mostrarle ni los precios ni la disponibilidad

forma del anticuerpo

purified antibody

tipo de anticuerpo

primary antibodies

clon

monoclonal

mol peso

(~27 kDa observed; 26.89 kDa calculated. Uncharacterized bands may be observed in some lysate(s).)

purificado por

using protein G

reactividad de especies

E. coli, mouse

isotipo

IgG1, kappa

Nº de acceso UniProt

Especificidad

Clone 2B6 is a mouse monoclonal antibody that detects GFP-conjugated to most proteins.

Aplicación

Anti-GFP, clone 2B6, Cat. No. MABC1689, is a mouse monoclonal antibody that detects GFP-conjugated to most proteins and has been tested for use in Immunocytochemistry and Western Blotting.
Immunocytochemistry Analysis: A 1:50 dilution from a representative lot detected GFP in HEK293T cells stabely transfected with GFP-tagged Aquaporin 4.
Western Blotting Immunocytochemistry

Características y beneficios

His-tagged recombinant jellyfish Green fluorescent protein.

Almacenamiento y estabilidad

Stable for 1 year at 2-8°C from date of receipt.

Nota de análisis

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Otras notas

Green fluorescent protein (UniProt: P42212) is encoded by the GFP gene in Aequorea victoria (Jellyfish). Green fluorescence protein (GFP) is a 27 kDa protein from jellyfish that serves as an energy-transfer acceptor. It transduces the blue chemiluminescence of the protein aequorin into green fluorescent light by energy transfer and it fluoresces in vivo upon receiving energy from the Ca2+-activated photoprotein aequorin. GFP has an absorption max at 395 nm and the emission peaks at 507 to 510 nm. However, the exact value of emission max depends on the environment of the chromophore. GFP is an important tool in cell biology research that displays stability under fixation conditions and is suitable for a variety of applications, such as tracing cell lineage tracer, reporter of gene expression, or exploring protein-protein interactions.
Evaluated by Western Blotting in lysates from HeLa cells transfected with GFP.

Western Blotting Analysis: 1 µg/mL of this antibody detected GFP in lysates from HeLa cells transfected with GFP.
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

¿No encuentra el producto adecuado?  

Pruebe nuestro Herramienta de selección de productos.


Certificados de análisis (COA)

Busque Certificados de análisis (COA) introduciendo el número de lote del producto. Los números de lote se encuentran en la etiqueta del producto después de las palabras «Lot» o «Batch»

¿Ya tiene este producto?

Encuentre la documentación para los productos que ha comprado recientemente en la Biblioteca de documentos.

Visite la Librería de documentos

Nuestro equipo de científicos tiene experiencia en todas las áreas de investigación: Ciencias de la vida, Ciencia de los materiales, Síntesis química, Cromatografía, Analítica y muchas otras.

Póngase en contacto con el Servicio técnico