Saltar al contenido
Merck
Todas las fotos(2)

Documentos clave

ABD16

Sigma-Aldrich

Anti-IKAROS Antibody

from rabbit, purified by affinity chromatography

Sinónimos:

DNA-binding protein Ikaros, Ikaros family zinc finger protein 1, Lymphoid transcription factor LyF-1

Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
En este momento no podemos mostrarle ni los precios ni la disponibilidad

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, human, mouse

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... IKZF1(10320)
mouse ... Ikzf1(22778)
rat ... Ikzf1(305501)

Categorías relacionadas

General description

IKAROS, also known as IKZF1, is a transcription factor that is important for the regulation of lymphopoiesis and also the development of the immune system. It has also been linked to adult and fetal erythroid differentiation in humans. IKAROS has been shown to induce local chromatin accessibility during CD4+ T cell differentiation and therefore, activating Th2 expression.

Immunogen

KLH-conjugated linear peptide corresponding to human IKAROS.

Application

Anti-IKAROS Antibody detects level of IKAROS & has been published & validated for use in WB & IC.
Immunocytochemistry Analysis: 1:500 dilution from a representative lot detected IKAROS in ReNcell human neural progenitor cells.
Research Category
Stem Cell Research
Research Sub Category
Transcription Factors

Quality

Evaluated by Western Blot in HeLa cell lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected IKAROS on 10 µg of HeLa cell lysate.

Target description

~65 kDa observed. (Martin-Ibanez, R., et al. (2010). The Journal of Comparative Neurology. 518:329–351.) An uncharacterized band appears at ~57 kDa in some lysates.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

¿No encuentra el producto adecuado?  

Pruebe nuestro Herramienta de selección de productos.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificados de análisis (COA)

Busque Certificados de análisis (COA) introduciendo el número de lote del producto. Los números de lote se encuentran en la etiqueta del producto después de las palabras «Lot» o «Batch»

¿Ya tiene este producto?

Encuentre la documentación para los productos que ha comprado recientemente en la Biblioteca de documentos.

Visite la Librería de documentos

Questions

Reviews

No rating value

Active Filters

Nuestro equipo de científicos tiene experiencia en todas las áreas de investigación: Ciencias de la vida, Ciencia de los materiales, Síntesis química, Cromatografía, Analítica y muchas otras.

Póngase en contacto con el Servicio técnico