96100025
Vantage L Adjuster Cartridge Housing Kit
Seleccione un Tamaño
EUR 106,00
Seleccione un Tamaño
About This Item
EUR 106,00
material
glass filled polypropylene surface (exterior)
Quality Level
manufacturer/tradename
Vantage
technique(s)
protein purification: suitable
diam.
2.2 cm (0.9 in.)
1 of 4
Este artículo | 96100029 | 96100032 | 96100030 |
---|---|---|---|
Vantage | Vantage | Vantage | Vantage |
protein purification: suitable | protein purification: suitable | protein purification: suitable | protein purification: suitable |
2.2 cm (0.9 in.) | 1.6 cm (0.6 in.) | 4.4 cm (1.7 in.) | 2.2 cm (0.9 in.) |
SP1 | SP1 | SP1 | SP1 |