1.67207
Reducer (PTFE) from S40 to S38
About This Item
Productos recomendados
storage temp.
no temp limit
General description
Special S40 bottle adapters made of PTFE provide all possible connection options for safe and reliable daily solvents handling. This adapter is equipped with S40 inner thread and GL45 outer thread.