Z675075
Stuart™ heat block for round bottom flasks
robust aluminum block transforms hotplates into heating mantles
About This Item
Código UNSPSC:
41102406
Materiales
aluminum
envase
pkg of 1 ea
fabricante / nombre comercial
Stuart SB16/6
Descripción general
This robust aluminum block transforms hotplates into heating mantles. Can be used with any hotplate that has 160 mm plate size. Four different sized holes for 25 mL, 50 mL, 100 mL and 250 mL round bottom flasks. Pre-drilled hole for thermometer or temperature probe.
- Accommodates 25 mL, 50 mL, 100 mL and 250 mL flasks
- Fits any 160 mm hotplate
- Aluminum allows fast consistent heat transfer
- Four feet secure block to hotplate
Información legal
Stuart is a trademark of Bibby Sterlin Ltd.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)