Documentos clave
Z645230
IKA® M 23 star-shaped cutting blade
M 23 star-shaped cutter, 1/cs
Seleccione un Tamaño
1 EA
US$ 853,00
US$ 853,00
PRECIOS SIN IMPUESTOS NACIONALES
Check Cart for Availability
Seleccione un Tamaño
Cambiar Vistas
1 EA
US$ 853,00
About This Item
Código UNSPSC:
41121800
NACRES:
NB.74
US$ 853,00
PRECIOS SIN IMPUESTOS NACIONALES
Check Cart for Availability
Materiales
stainless steel
envase
case of 1
pack of 1
fabricante / nombre comercial
IKA 1443400
idoneidad
suitable for
Descripción general
stainless steeel. star knife for use with the IKA M 20 Universal Mill. For shredding fibrous materials such as paper and plant material, but also for plastics and other materials with a low specific weight.
used to crush fibrous substances such as paper and vegetation, but also for plastics and material with a low specific weight.
Información legal
IKA is a registered trademark of IKA-Werke GmbH & Co. KG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)