Documentos clave
Z564265
Vacuum accessories for Buchi® vacuum pumps
Secondary condenser
Seleccione un Tamaño
1 EA
US$ 3.860,00
US$ 3.860,00
PRECIOS SIN IMPUESTOS NACIONALES
Check Cart for Availability
Seleccione un Tamaño
Cambiar Vistas
1 EA
US$ 3.860,00
About This Item
Código UNSPSC:
41105101
NACRES:
NB.43
US$ 3.860,00
PRECIOS SIN IMPUESTOS NACIONALES
Check Cart for Availability
envase
pack of 1 ea
fabricante / nombre comercial
Büchi 47180
Descripción general
This compact high performance condenser produces maximum condensation of residual solvent vapor. It also separates any liquid solvent from the pump, aputting it directly into the receiving flask. Insulation sleeves prevent the formation of undesirable condensation and provide the condenser with effective protection against mechanical damage.
Información legal
Büchi is a registered trademark of Büchi Labortechnik AG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Visite la Librería de documentos
Questions
  1. Per il prodotto Z564265 il condensatore secondario è raffreddato con flusso d'acqua oppure con Co2 solida?
    1 answer
    1. This secondary condenser is used with recirculating water or coolant. This product does not have the cold trap condenser.