Documentos clave
Z531774
Sigma-Aldrich
Tubing Adapters with Torion Connection
O.D. 5/16 in., With hose barb, male joint: ST/NS 24/40
Seleccione un Tamaño
1 EA
US$ 209,00
US$ 209,00
PRECIOS SIN IMPUESTOS NACIONALES
Check Cart for Availability
Seleccione un Tamaño
Cambiar Vistas
1 EA
US$ 209,00
About This Item
Código UNSPSC:
41121815
NACRES:
NB.35
US$ 209,00
PRECIOS SIN IMPUESTOS NACIONALES
Check Cart for Availability
Junta
male joint: ST/NS24/40
D.E.
5/16 in.
7.9 mm
Descripción general
The unique Torion connection ensures an airtight seal between tubing and reaction vessels when transferring air-sensitive materials.
5/16 in. O.D fits a #8 Torion, 1/2 in. O.D. fits a #14 Torion, and 3/16 in. O.D fits a #5 Torion.
  • Hard-wall transfer tubing (e.g. PP or PTFE) is inserted through the threaded polypropylene bushing and PTFE-coated sealing ring of the adapter
  • Turn the bushing to make an airtight compression seal with the tubing
  • Use the optional solid-glass plug to make an airtight Torion seal when the transfer tubing is not in use
  • O.D. refers to the tubing outer diameter
Información legal
Torion is a trademark of Bibby Sterlin Ltd.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)