Documentos clave
Z300934
Quickfit® Reduction Adapters
top female joint: ST/NS 19/26, bottom male joint: ST/NS 34/35
Sinónimos:
DA25
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
En este momento no podemos mostrarle ni los precios ni la disponibilidad
joint
female joint: ST/NS top19/26
male joint: ST/NS bottom34/35
manufacturer/tradename
Quickfit DA 25
availability
not available in USA and Canada
General description
These adapters connect items with a smaller cone to a piece of glassware with a larger size socket.
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)