Z250902
InsulEze® insulating kit
Combination
About This Item
Código UNSPSC:
41112303
NACRES:
NB.76
Productos recomendados
fabricante / nombre comercial
Glas-Col 100GIK240
Descripción general
Includes one each of kits A, B, and C.
Make insulating less time-consuming, awkward, and unsightly in the lab. Use for heat or cold shielding, insulating over heaters, or as shock absorber. Insulating materials can be cut and shaped to almost any size with minimum fraying of the edges. Material is non-hazardous, non-flammable, and inert to most chemicals.
Información legal
InsulEze is a registered trademark of Glas-Col Laboratory Products
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)